ENA369381634 Enamine

7-chloroquinoline-2-carboxamide

  • MDL number MFCD16038707
  • Cas Number : 65147-97-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100518

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100518
Availability details for
Quantity
  • Availability
  • Add to Cart