ENA369381636 Enamine

2-[2-(1-methoxyethyl)-1,3-thiazol-4-yl]acetic acid

  • MDL number MFCD21938391
  • Cas Number : 1376100-18-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100066

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100066
Availability details for
Quantity
  • Availability
  • Add to Cart