ENA369381640 Enamine

2-(methoxycarbonyl)pyridine-4-carboxylic acid

  • MDL number MFCD00661398
  • Cas Number : 24195-10-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 228...230
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100069

Safety Information

msds https://www.enaminestore.com/catalog/EN300-100069
Availability details for
Quantity
  • Availability
  • Add to Cart