ENA369381652 Enamine

7-amino-2,3-dihydro-1,4-benzodioxine-5-carboxylic acid

  • MDL number MFCD17276600
  • Cas Number : 66410-92-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100578

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100578
Availability details for
Quantity
  • Availability
  • Add to Cart