ENA369381656 Enamine

2-amino-3-methoxy-2-methylpropanoic acid hydrochloride

  • MDL number MFCD22056367
  • Cas Number : 954120-52-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100589

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100589
Availability details for
Quantity
  • Availability
  • Add to Cart