ENA369381663 Enamine

2-amino-3-methoxy-2-methylpropan-1-ol

  • MDL number MFCD16788617
  • Cas Number : 96388-72-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100596

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100596
Availability details for
Quantity
  • Availability
  • Add to Cart