ENA369381664 Enamine

1-(2-phenylethyl)-1H-pyrazole

  • MDL number MFCD18889180
  • Cas Number : 58416-69-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100597

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-100597
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart