ENA369381670 Enamine

1-methyl-2,3-dihydro-1H-inden-2-one

  • MDL number MFCD22196376
  • Cas Number : 35587-60-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100606

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100606
Availability details for
Quantity
  • Availability
  • Add to Cart