ENA369381679 Enamine

4-(2-fluorophenyl)-1,2,5-oxadiazol-3-amine

  • MDL number MFCD11574182
  • Cas Number : 21742-05-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100706

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100706
Availability details for
Quantity
  • Availability
  • Add to Cart