ENA369381681 Enamine

1-(5-ethyl-1,3-thiazol-2-yl)ethan-1-amine

  • MDL number MFCD09904508
  • Cas Number : 1017391-90-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100709

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100709
Availability details for
Quantity
  • Availability
  • Add to Cart