ENA369381685 Enamine

3-[3-(1-methoxyethyl)-1,2,4-oxadiazol-5-yl]propanoic acid

  • MDL number MFCD21938390
  • Cas Number : 1376269-03-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100071

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100071
Availability details for
Quantity
  • Availability
  • Add to Cart