ENA369381686 Enamine

2-(2-bromoethyl)-5-phenyl-2H-1,2,3,4-tetrazole

  • MDL number MFCD11129317
  • Cas Number : 1019340-69-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100718

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100718
Availability details for
Quantity
  • Availability
  • Add to Cart