ENA369381687 Enamine

4-amino-3-ethoxy-N,N-dimethylbenzamide

  • MDL number MFCD18338473
  • Cas Number : 1343088-57-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-100719

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100719
Availability details for
Quantity
  • Availability
  • Add to Cart