ENA369381692 Enamine

1-(3-fluorophenyl)-2,2-dimethylpropan-1-amine

  • MDL number MFCD12154114
  • Cas Number : 681514-01-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100724

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100724
Availability details for
Quantity
  • Availability
  • Add to Cart