ENA369381693 Enamine

1-(3-bromo-5-fluorophenyl)propan-2-ol

  • MDL number MFCD22056371
  • Cas Number : 1376240-04-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100725

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100725
Availability details for
Quantity
  • Availability
  • Add to Cart