ENA369381701 Enamine

5-chloro-4,6-dimethyl-1,3-benzoxazol-2-amine

  • MDL number MFCD16742790
  • Cas Number : 1248171-77-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100770

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100770
Availability details for
Quantity
  • Availability
  • Add to Cart