ENA369381704 Enamine

4-(hydroxymethyl)-1-(2-phenylethyl)pyrrolidin-2-one

  • MDL number MFCD10003998
  • Cas Number : 904237-82-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100773

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100773
Availability details for
Quantity
  • Availability
  • Add to Cart