ENA369381705 Enamine

2-amino-2-(5-bromo-2-methoxyphenyl)ethan-1-ol

  • MDL number MFCD12827200
  • Cas Number : 1178727-53-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 99...101
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100774

Safety Information

msds https://www.enaminestore.com/catalog/EN300-100774
Availability details for
Quantity
  • Availability
  • Add to Cart