ENA369381711 Enamine

2-(1H-imidazol-1-yl)-1,3-benzothiazol-6-amine

  • MDL number MFCD18298710
  • Cas Number : 1341887-20-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100781

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100781
Availability details for
Quantity
  • Availability
  • Add to Cart