ENA369381719 Enamine

5-amino-2-fluoro-N,N-dimethylbenzamide

  • MDL number MFCD12187396
  • Cas Number : 682757-75-1
  • Molecular Weight : 182.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-100794

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100794
Availability details for
Quantity
  • Availability
  • Add to Cart