ENA369381735 Enamine

3-(4-fluorophenyl)-3-hydroxypropanoic acid

  • MDL number MFCD16302337
  • Cas Number : 40620-62-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100833

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100833
Availability details for
Quantity
  • Availability
  • Add to Cart