ENA369381739 Enamine

1-(quinolin-2-yl)ethan-1-one

  • MDL number MFCD08062664
  • Cas Number : 1011-47-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 47...49
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100843

Safety Information

msds https://www.enaminestore.com/catalog/EN300-100843
Availability details for
Quantity
  • Availability
  • Add to Cart