ENA369381747 Enamine

4-(phenylmethylidene)piperidine

  • MDL number MFCD20664129
  • Cas Number : 164650-58-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100855

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100855
Availability details for
Quantity
  • Availability
  • Add to Cart