ENA369381750 Enamine

(3S)-3-amino-3-(4-chlorophenyl)propanamide

  • MDL number MFCD12797265
  • Cas Number : 1308301-15-6
  • Molecular Weight : 198.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3S)-3-amino-3-(4-chlorophenyl)propanamide
Inchi Code 1S/C9H11ClN2O/c10-7-3-1-6(2-4-7)8(11)5-9(12)13/h1-4,8H,5,11H2,(H2,12,13)/t8-/m0/s1
Inchi Key HUEMYQOYOXHOJL-QMMMGPOBSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-100858

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100858
Availability details for
Quantity
  • Availability
  • Add to Cart