ENA369381751 Enamine

3-[(1S)-1-hydroxyethyl]benzene-1-sulfonamide

  • MDL number MFCD18842663
  • Cas Number : 1372452-60-2
  • Molecular Weight : 201.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100859

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100859
Availability details for
Quantity
  • Availability
  • Add to Cart