ENA369381753 Enamine

2-(2-methylphenoxy)butanoic acid

  • MDL number MFCD03422242
  • Cas Number : 161790-50-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100861

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100861
Availability details for
Quantity
  • Availability
  • Add to Cart