ENA369381755 Enamine

N-methyl-2-(pyrrolidin-1-yl)pyridin-3-amine

  • MDL number MFCD17291856
  • Cas Number : 1341659-86-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105...107
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100081

Safety Information

msds https://www.enaminestore.com/catalog/EN300-100081
Availability details for
Quantity
  • Availability
  • Add to Cart