ENA369381759 Enamine

3-amino-4-methoxy-3-methylbutan-1-ol

  • MDL number MFCD21184408
  • Cas Number : 1376105-62-2
  • Molecular Weight : 133.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-amino-4-methoxy-3-methyl-1-butanol
Inchi Code 1S/C6H15NO2/c1-6(7,3-4-8)5-9-2/h8H,3-5,7H2,1-2H3
Inchi Key OSKKFJANHDTOGL-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-100870

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100870
Availability details for
Quantity
  • Availability
  • Add to Cart