ENA369381762 Enamine

1-cyclopropyl-1,4-diazepan-5-one

  • MDL number MFCD22056380
  • Cas Number : 30858-70-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98...100
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100873

Safety Information

msds https://www.enaminestore.com/catalog/EN300-100873
Availability details for
Quantity
  • Availability
  • Add to Cart