ENA369381763 Enamine

2-(difluoromethoxy)-4-fluorobenzoic acid

  • MDL number MFCD14698509
  • Cas Number : 1214347-60-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115...117
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100874

Safety Information

msds https://www.enaminestore.com/catalog/EN300-100874
Availability details for
Quantity
  • Availability
  • Add to Cart