ENA369381764 Enamine

4-chloro-N-(hydroxymethyl)benzamide

  • MDL number MFCD00558390
  • Cas Number : 13855-74-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 145...147
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100875

Safety Information

msds https://www.enaminestore.com/catalog/EN300-100875
Availability details for
Quantity
  • Availability
  • Add to Cart