ENA369381765 Enamine

1-(2,6-dimethylpyrimidin-4-yl)piperidin-4-amine

  • MDL number MFCD07365290
  • Cas Number : 893754-92-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100877

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100877
Availability details for
Quantity
  • Availability
  • Add to Cart