ENA369381768 Enamine

2,2,2-trifluoro-N-(4-hydroxycyclohexyl)acetamide

  • MDL number MFCD12819946
  • Cas Number : 1183415-13-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100880

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100880
Availability details for
Quantity
  • Availability
  • Add to Cart