ENA369381774 Enamine

2-(2H-1,2,3-triazol-2-yl)acetic acid

  • MDL number MFCD03840810
  • Cas Number : 4320-91-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100916

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100916
Availability details for
Quantity
  • Availability
  • Add to Cart