ENA369381775 Enamine

4-methyl-2-oxo-2H-chromene-3-carboxylic acid

  • MDL number MFCD06208395
  • Cas Number : 833-31-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 163...165
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100917

Safety Information

msds https://www.enaminestore.com/catalog/EN300-100917
Availability details for
Quantity
  • Availability
  • Add to Cart