ENA369381777 Enamine

3-methyl-5-phenylcyclohexan-1-one

  • MDL number MFCD19301755
  • Cas Number : 60741-75-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-100920

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100920
Availability details for
Quantity
  • Availability
  • Add to Cart