ENA369381779 Enamine

4-(decylamino)butanoic acid hydrochloride

  • MDL number MFCD22056330
  • Cas Number : 1376391-99-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100088

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100088
Availability details for
Quantity
  • Availability
  • Add to Cart