ENA369381781 Enamine

6-(methylamino)-2,3,4,5-tetrahydro-1,2,4-triazine-3,5-dione

  • MDL number MFCD00703053
  • Cas Number : 64842-42-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100927

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100927
Availability details for
Quantity
  • Availability
  • Add to Cart