ENA369381782 Enamine

6-chloro-N4-cyclopropylpyrimidine-4,5-diamine

  • MDL number MFCD09743305
  • Cas Number : 195252-62-1
  • Molecular Weight : 184.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-chloro-N~4~-cyclopropyl-4,5-pyrimidinediamine
Inchi Code 1S/C7H9ClN4/c8-6-5(9)7(11-3-10-6)12-4-1-2-4/h3-4H,1-2,9H2,(H,10,11,12)
Inchi Key UOZWNOIHCDKJPY-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-100928

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100928
Availability details for
Quantity
  • Availability
  • Add to Cart