ENA369381786 Enamine

N-(2,2,2-trifluoroethyl)cyclobutanamine hydrochloride

  • MDL number MFCD22056387
  • Cas Number : 1116589-04-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 229...231
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100947

Safety Information

msds https://www.enaminestore.com/catalog/EN300-100947
Availability details for
Quantity
  • Availability
  • Add to Cart