ENA369381792 Enamine

benzyl N-(1H-pyrazol-4-ylmethyl)carbamate

  • MDL number MFCD12850715
  • Cas Number : 1184024-26-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 126...128
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100955

Safety Information

msds https://www.enaminestore.com/catalog/EN300-100955
Availability details for
Quantity
  • Availability
  • Add to Cart