ENA369381794 Enamine

tert-butyl N-(6-amino-2H-1,3-benzodioxol-5-yl)carbamate

  • MDL number MFCD12432277
  • Cas Number : 1154989-54-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 93...95
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100957

Safety Information

msds https://www.enaminestore.com/catalog/EN300-100957
Availability details for
Quantity
  • Availability
  • Add to Cart