ENA369381796 Enamine

methyl 2-amino-3-(6-fluoro-1H-indol-3-yl)propanoate hydrochloride

  • MDL number MFCD22056390
  • Cas Number : 1018815-57-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100959

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100959
Availability details for
Quantity
  • Availability
  • Add to Cart