ENA369381797 Enamine

4-(methylsulfanyl)pyridin-3-amine

  • MDL number MFCD11635404
  • Cas Number : 38240-24-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 78...80
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100961

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-100961
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart