ENA369381799 Enamine

2-(aminomethyl)-5-hydroxy-1-methyl-1,4-dihydropyridin-4-one dihydrobromide

  • MDL number MFCD22056391
  • Cas Number : 1376050-72-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 249...251
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100963

Safety Information

msds https://www.enaminestore.com/catalog/EN300-100963
Availability details for
Quantity
  • Availability
  • Add to Cart