ENA369381801 Enamine

(3R)-3-amino-3-(3-chlorophenyl)propanamide hydrochloride

  • MDL number MFCD22056393
  • Cas Number : 1376000-87-1
  • Molecular Weight : 235.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3R)-3-amino-3-(3-chlorophenyl)propanamide hydrochloride
Inchi Code 1S/C9H11ClN2O.ClH/c10-7-3-1-2-6(4-7)8(11)5-9(12)13;/h1-4,8H,5,11H2,(H2,12,13);1H/t8-;/m1./s1
Inchi Key RZLRCKQKMJXBTQ-DDWIOCJRSA-N

Description

Purity 95
Salt Data Cl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-100969

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100969
Availability details for
Quantity
  • Availability
  • Add to Cart