ENA369381814 Enamine

4-amino-1-methyl-2,5-dihydro-1H-imidazol-2-one

  • MDL number MFCD19215909
  • Cas Number : 1190594-65-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101037

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-101037
Availability details for
Quantity
  • Availability
  • Add to Cart