ENA369381819 Enamine

8-azabicyclo[5.2.0]nonan-9-one

  • MDL number MFCD19217966
  • Cas Number : 96850-68-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101043

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-101043
Availability details for
Quantity
  • Availability
  • Add to Cart