ENA369381829 Enamine

2-(2-phenylethyl)cyclopentan-1-amine

  • MDL number MFCD16824901
  • Cas Number : 63762-62-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Product Link https://www.enaminestore.com/catalog/EN300-100103

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100103
Availability details for
Quantity
  • Availability
  • Add to Cart