ENA369381831 Enamine

4-(2-methylphenyl)-1,2,5-oxadiazol-3-amine

  • MDL number MFCD16779602
  • Cas Number : 22899-59-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-101063

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-101063
Availability details for
Quantity
  • Availability
  • Add to Cart